Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR® Talon®, Stand plate, rectangular

VWR® Talon®, Stand plate, rectangular

Support stand base, rectangular, 127×203 mm
Catalog # VWRU12985-070
Supplier:   VWR Collection
VWR® Talon®, Stand plate, rectangular
Catalog # VWRU12985-070
Supplier:  VWR Collection
VWR® Talon®, Stand plate, rectangular
VWR® Talon®, Stand plate, rectangular

Support stand base, rectangular, 127×203 mm

Price
₩242,910
Log in for availability
1 * 1 items
Product Details & Documents

Cast iron with black enamel finish.

  • All bases feature a built-in support rod holder with locking knob that accepts a 13 mm Ø support rod (sold separately)
  • For use with VWR® Talon® lab frames
Specifications
  • Description:
    Support stand base, rectangular
  • L×W:
    127×203 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.