₩0
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Korea, Republic of
ContactUsLinkComponent
10% Carbon dioxide CO2 in Nitrogen N2for blood gas analysis.Empty/Unwashed 1 * 1.280
605003N.EMPTY
:
VWR Chemicals
10% Carbon dioxide CO2 in Nitrogen N2for blood gas analysis.Empty/Unwashed 1 * 1.280
605003N.EMPTY
:
VWR Chemicals
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection