Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
Dichloromethane ≥98% stabilised, TECHNICAL

Dichloromethane ≥98% stabilised, TECHNICAL

Metal drum, 190 L
Catalog # 25629.557
Health
Supplier:   VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Catalog # 25629.557
Supplier:  VWR Chemicals
CAS Number:  75-09-2
Dichloromethane ≥98% stabilised, TECHNICAL
Dichloromethane ≥98% stabilised, TECHNICAL

Metal drum, 190 L

CAS Number:  75-09-2
Price
Inquire For Price
Log in for availability
1 * 190 L
Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilised with ethanol 0.15 %
  • Storage Temperature:Ambient
  • UN:1593
  • REACH:01-2119480404-41
Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 98 %

Specifications

  • Pack type:
    Metal drum
  • Pk:
    190 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.