Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Funnels, Analytical
VWR®, Funnels, Analytical
VWR®, Funnels, Analytical
Supplier:  VWR Collection
Certificates
VWR®, Funnels, Analytical
VWR®, Funnels, Analytical
16 Options Available Below
Product Details & Documents
PP, translucent.
  • Inner ribs at an angle of 58° for rapid filtration
  • Outer ribs prevent air from being trapped under the filter paper
  • Very good chemical and temperature resistance
Designed for use with standard filter paper. Cone angle 60°.
Product Family Options
Product Information
  • Length
    Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 75,8 mm
    49,3 mm
    6,3 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
  • 75,8 mm
    49,3 mm
    6,3 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - ₩77,000
    ₩77,000
    1 * 12 items
       ₩77,000
  • 84,6 mm
    52,0 mm
    7,1 mm
    45 mm
    Log in for availability
    Log in for availability
  • 84,6 mm
    52,0 mm
    7,1 mm
    45 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - ₩82,805
    ₩82,805
    1 * 12 items
       ₩82,805
  • 103,4 mm
    59,5 mm
    7,1 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
  • 103,4 mm
    59,5 mm
    7,1 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - ₩87,710
    ₩87,710
    1 * 12 items
       ₩87,710
  • 111,7 mm
    61,4 mm
    6,4 mm
    65 mm
    Log in for availability
    Log in for availability
  • 111,7 mm
    61,4 mm
    6,4 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 111,7 mm
    61,4 mm
    6,4 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - ₩99,621
    ₩99,621
    1 * 12 items
       ₩99,621
  • 139,4 mm
    76,3 mm
    6,8 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
  • 139,4 mm
    76,3 mm
    6,8 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - ₩70,408
    ₩70,408
    1 * 6 items
       ₩70,408
  • 164,0 mm
    89,0 mm
    8,2 mm
    90 mm
    Log in for availability
    Log in for availability
  • 164,0 mm
    89,0 mm
    8,2 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - ₩83,031
    ₩83,031
    1 * 6 items
       ₩83,031
  • 180,0 mm
    95,4 mm
    9,1 mm
    100 mm
    Log in for availability
    Log in for availability
  • 180,0 mm
    95,4 mm
    9,1 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items - ₩67,487
    ₩67,487
    1 * 4 items
       ₩67,487
  • 280,0 mm
    154,0 mm
    14,0 mm
    160 mm