

Copper(II) sulphate pentahydrate 99.0-101.0%, crystallised Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7758-99-8
:
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681
:249.69 g/mol
Merck Index:13,02682
Density:2.284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-847-6
UN:3077
ADR:9,III
REACH:01-2119520566-40
:GHS07
:GHS09
Specification Test Results
- Assay:99.0 - 101.0 %
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification (SO4):Passes test
- Appearance of solution:Passes test
- Cl (Chloride):≤ 100 ppm
- Fe (Iron):≤ 100 ppm
- Loss on drying (250 ± 10°C):35.0 - 36.5 %
- Residual solvents:Unlikely by manuf.process
- Residual metal catalysts (skip lot)::.
- As (Arsenic):≤ 2 ppm
- Cd (Cadmium):≤ 1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 2 ppm
- Ni (Nickel):≤ 3 ppm
- Pb (Lead):≤ 2 ppm
- V (Vanadium):≤ 2 ppm
- Conforms to Ph.Eur.:Passes test
Specification Test Results
Frequently Bought Together