


Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99%, Biotechnology Grade
제품 번호 0883-100ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless, 2-layer liquid
- DNase:none detected
- O.D.@ 330nm:= 0.2
- O.D.@ 405nm:= 0.1
- O.D.@ 510nm:= 0.1
- pH @25°C:6.5 - 6.9
- Protease:none detected
- Purity:= 99%
- RNase:none detected
Specification Test Results
About this item
:
UN:2810
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Storage Temperature:Refrigerator
CAS Number:136112-00-0
MDL Number:MFCD00133763