VWR®, Tris-borate-EDTA (TBE) buffer
TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose. Buffers are available as liquid concentrate or Ready-Pack™ (a foil pouch containing sufficient material to prepare 1 litre of 10X concentrate).
- High buffering capacity
- High ionic strength
- Allows extended electrophoresis runs without buffer recirculation
- Convenient packaging options minimise weighing and mixing
- Free from DNase, RNase and protease
Composition of (1X) solution contains 0,089 M Tris base, 0,089 M borate and 0,002 M EDTA, disodium salt, dihydrate with a final pH of 8,3.
: *Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.
TBE is optimised for DNA electrophoresis techniques using both acrylamide and agarose. Buffers are available as liquid concentrate or Ready-Pack™ (a foil pouch containing sufficient material to prepare 1 litre of 10X concentrate).
- High buffering capacity
- High ionic strength
- Allows extended electrophoresis runs without buffer recirculation
- Convenient packaging options minimise weighing and mixing
- Free from DNase, RNase and protease
Composition of (1X) solution contains 0,089 M Tris base, 0,089 M borate and 0,002 M EDTA, disodium salt, dihydrate with a final pH of 8,3.
: *Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.
Product Family Options
- DescriptionPack typePk
- TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapurePlastic bottle1 L1 * 1 L