VWR®, Funnels with Long Stem
공급사: VWR Collection
Jump to:
이 제품에 관하여
PTFE, white.
- Temperature resistance: –200 to +280 °C
- Totally chemically inert
- Super smooth non-stick internal finish
Conical opening with long stem.
5 Options Available Below
Product Details & Documents
PTFE, white.
- Temperature resistance: –200 to +280 °C
- Totally chemically inert
- Super smooth non-stick internal finish
Conical opening with long stem.
Product Family Options
제품 정보
- LengthStem lengthStem O-ØFunnel O-Ø재고 여부Price