Order Entry
Korea, Republic of
ContactUsLinkComponent
 
Petroleum spirit, 20…40 °C, BAKER ANALYZED®, J.T.Baker®
Catalog # 9272-03
Supplier:  Avantor
CAS Number:  
Petroleum spirit, 20…40 °C, BAKER ANALYZED®, J.T.Baker®
Catalog # 9272-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Appearance and Color:
    Passes Test
  • Residue after Evaporation:
    ≤0.0020 %
  • Acidity:
    Passes Test
  • Heavy Oils and Fats:
    Passes Test
  • Boiling Range(Lower):
    20 - 40 °C
  • Boiling Range(Upper):
    20 - 40 °C

Specification Test Results

About this item

 :
Boiling Pt:20… 40 °C (1013 hPa)
Density:0.62 g/cm³ (20 °C)
ADR:3,I
 :Management of Change (MOC) category = N
CAS Number:64742-49-0
 :8008-20-6
MDL Number:MFCD00081849