VWR®, Digital Stopwatch
Catalog # 609-0054
공급사: VWR Collection
사양
- Colour:Yellow
- 설명:Digital stopwatch
- Seq:01
- W×D×H:54×60×12,7 mm
- Pk:1
사양
이 제품에 관하여
O-ring seal and shockproof plastic case for lab and plant use.
- Timing capacity: 23 hours, 59 minutes, 59 seconds (count up)
- LCD shows time of day in hours, minutes and seconds
- AM/PM, date and day of the week
- Cumulative split timing
- Audible alarm sounds for 1 minute
- Waterproof to a pressure of three atmospheres