Order Entry
Korea, Republic of
ContactUsLinkComponent
Water, Electran for molecular biology
Water, Electran for molecular biology
Catalog # 445847D
Supplier:  VWR Chemicals
CAS Number:  
undefined
Water, Electran for molecular biology
Catalog # 445847D
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Osmolality (mOsm/kg):
    Max. 3
  • DNases:
    Not detected
  • RNases:
    Not detected
  • Proteases:
    Not detected
  • Fe (Iron):
    Max. 0.006 ppm
  • Mg (Magnesium):
    Max. 0.005 ppm
  • Pb (Lead):
    Max. 0.005 ppm

Specification Test Results

About this item

 :
 :18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332