Water nuclease-free
Catalog # 436912C
Supplier: VWR Chemicals
CAS Number:
Specifications
- 포장 유형:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Osmolality (mOsm/kg):Max. 3
- DNases:Not detected
- RNases:Not detected
- Proteases:Not detected
Specification Test Results
About this item
:
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:0000
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332