



Cyclohexane ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
:110-82-7
:
:C₆H₁₂
:MFCD00003814
:84.16 g/mol
:13,02752
:81 °C (1013 hPa)
:7 °C
:0.7785 g/cm³ (20 °C)
:–18 °C
:203-806-2
:1145
:3,II
:01-2119463273-41
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
- Assay (on anhydrous substance):Min. 99.5 %
- IR Spectrum:Passes test
- Readily carbonisable substances:Passes test
- Boiling point:79 - 81 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.778 - 0.779
- Density (20/20):0.779 - 0.780
- Free acidity:Max. 0.0003 meq/g
- Solidification point:5.5 - 6.5 °C
- Substances coloured by H2SO4:Max. 60 APHA
- Benzene:Max. 100 ppm
- Cyclohexene:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.1 ppm
- Ba (Barium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0,06 ppm
- Na (Sodium):Max. 1 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Packaging Information
spec_VWRC23224.293 spec_VWRC23224.327 spec_VWRC23224.362 spec_VWRC23224.464 spec_VWRC23224.550
Product specification
More
Frequently Bought Together