VWR®, Funnels with Long Stem
Supplier: VWR Collection
Jump to:
About this item
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Smooth internal walls.
Certifications: ISO 4798.
3 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Smooth internal walls.
Certifications: ISO 4798.
Documents
coc_VWRI221-0172_GL1223 coc_VWRI221-0172_GL2724 coc_VWRI221-0172_GL2725 coc_VWRI221-0172_GL3722 coc_VWRI221-0172_GL4025
More
TDS VWR®, Funnels with Long Stem.pdf
Datasheet
TDS_VWR_COLLECTION_FUNNELS_LONG_STEM_BOROSILICATE_GLASS.pdf
Product specification
VWR® for filtration VWR® for rotary evaporation VWR® Glassware Solutions
Product Literature
Product Family Options
Product Information
- Stem lengthStem O-ØFunnel O-ØAvailabilityPrice