Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Funnels with Short Stem
VWR®, Funnels with Short Stem
VWR®, Funnels with Short Stem
Supplier:  VWR Collection
Certificates
VWR®, Funnels with Short Stem
VWR®, Funnels with Short Stem
8 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Smooth internal walls.
Certifications: ISO 4798.
Product Family Options
Product Information