Order Entry
Korea, Republic of
ContactUsLinkComponent
Hydrochloric acid 5.0 N, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Hydrochloric acid 5.0 N, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Irritant
Hydrochloric acid 5.0 N, Biotech Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    Passes test
  • CAUTION: For Manufacturing, processing or repackaging:
    4.98 - 5.02
  • Appearance (Clear, colorless liquid):
    Passes Test
  • Normality, eq/L:
    4.98 - 5.02
  • Sulfate (SO₄):
    ≤1 ppm
  • Heavy Metals (as Pb):
    ≤0.5 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Ammonium (NH₄):
    ≤3 ppm

Specification Test Results

Related Information

Frequently Bought Together