Sodium dihydrogen phosphate monohydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
:10049-21-5
:
:NaH₂PO₄·H₂O
:MFCD00149208
:137.99 g/mol
:13,08734
:100 °C
:2.04 g/cm³ (20 °C)
:Ambient
:231-449-2
:01-2119489796-13
- Assay:99.0 - 102.0 %
- Solution in water:Passes test
- pH (5 %):4.1 - 4.5
- Cl (Chloride):Max. 0.0005 %
- SO4 (Sulphate):Max. 0.005 %
- Total N (Nitrogen):Max. 0.001 %
- Heavy metals (as Pb):Max. 0.0005 %
- As (Arsenic):Max. 0.00005 %
- Ca (Calcium):Max. 0.005 %
- Cu (Copper):Max. 0.0002 %
- Fe (Iron):Max. 0.0005 %
- K (Potassium):Max. 0.005 %
- Pb (Lead):Max. 0.0005 %
Frequently Bought Together