Order Entry
Korea, Republic of
ContactUsLinkComponent
Orthophosphoric acid ≥75%, GPR RECTAPUR®
Corrosive
Orthophosphoric acid ≥75%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Orthophosphoric acid ≥75%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    75 - 78 %
  • Density (20/4):
    1.579 - 1.610
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together