VWR® Maximum SF Boot Covers, Fluid Impervious
About this item
VWR® Maximum SF boot covers are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) with breathable polyethylene film coating, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Non skid and anti-static
- Fluid impervious, sealed seams to repel water
- Superior friction coefficient to ensure maximum traction
- Maximum skid resistance on wet and smooth surfaces
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
* One size fits all
3 Options Available Below
Product Details & Documents
VWR® Maximum SF boot covers are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) with breathable polyethylene film coating, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Non skid and anti-static
- Fluid impervious, sealed seams to repel water
- Superior friction coefficient to ensure maximum traction
- Maximum skid resistance on wet and smooth surfaces
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
* One size fits all
Specifications for Product Family
Specifications
- Catalog No:
- 113-1185
- 113-1186
- 113-1184
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
Specifications
Product Family Options
Product Information
- ColourShoe sizeLengthAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-1185Garment applicationCleanroom|Laboratory - Specifications:
More
Specifications
Cat. No.113-1186Garment applicationCleanroom|Laboratory - Specifications:
More
Specifications
Cat. No.113-1184Garment applicationCleanroom|Laboratory