VWR® Aquaresist Water Bath Protective Agent
Prevents the growth of algae and bacteria in baths and circulating thermostats, for optimally hygienic working conditions.
- Durable: Effective for weeks, as displayed by a colour indicator
- Economical: Low consumption - only 1 ml solution per 1 litre of water
- Available with and without colour indicator
: Use biocides safely. Always read the label and product information before use.
Frequently Bought Together