Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Combined pH Electrodes, pHASE

VWR®, Combined pH Electrodes, pHASE

VWR®, Combined pH Electrodes, pHASE
Supplier:  VWR Collection
Certificates
VWR®, Combined pH Electrodes, pHASE
VWR®, Combined pH Electrodes, pHASE

5 Options Available Below

Product Details & Documents

Combined electrodes providing fast, accurate and stable pH measurements, ideal for control quality and research and development use.

  • Fast, accurate and stable pH measurements, fast thermal equilibration, high accuracy even in samples with large temperature range
  • Available with sealed design (economical, easy to use, with minimum maintenance) or Rapid-Renew design (refillable electrodes used with dirty or other samples that cause junction fouling)
  • Available with epoxy body (with good chemical resistance, maximum ruggedness and removable bulb guard) or glass body (maximum chemical resistance with fully exposed ruggedised bulbs)
  • With 1 m fixed cable

pH range: 0 to 14

Accuracy: to 0,04 pH between 0 to 100 °C in pH 7 buffer

Stability: 0,005 pH or less drift per hour

Temperature range: 0 to +100 °C

Temperature range: 0 to +100 °C

Product Family Options

Product Information

  • Type
    pH
    Material
    Electrolyte
    Ø×L
    Connection
    Length
    For
    Availability
    Price
  • Refillable, double junction (plastic)
    0 - 14
    Epoxy
    3,5 mol/l KCl
    12×150 mm
    BNC
    Most applications
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • Refillable, double junction (plastic)
    0 - 14
    Glass
    3,5 mol/l KCl
    12×150 mm
    BNC
    Aqueous media, beverage, Tris buffer, cosmetics, ultra-pure water
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • Sealed, double junction (Pellon)
    0 - 14
    Epoxy
    Gel
    12×150 mm
    BNC
    Aqueous media, beverage, Tris buffer
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • Sealed, double junction (Pellon)
    0 - 14
    Glass
    Gel
    12×150 mm
    BNC
    Aqueous media, beverage, Tris buffer
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

  • Sealed, flat tip, double junction (HDPE)
    0 - 14
    Epoxy
    Gel
    12×150 mm
    BNC
    150 mm
    Aqueous media, beverage, Tris buffer, cosmetics, ultra-pure water
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items

Accessories

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Refill solution 3,5 mol/l KCl
    Refill solution 3,5 mol/l KCl
    662-1784
    100 mL
    Log in for availability
    1 * 100 mL