VWR®, Weighing Boats
Supplier: VWR Collection
Jump to:
About this item
Borosilicate 3.3 glass.
- Ideal for weighing small samples
- Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
3 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass.
- Ideal for weighing small samples
- Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
Documents
Product Family Options
Product Information
- CapacityLengthAvailabilityPrice