VWR®, Pipette Tips, Standard
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
: Supplied loose or in racks holding 96 tips.
These universal tips are compatible with most micropipettes.
- Non sterile, autoclavable
- Smooth inside surface finish for minimum retention of dispensed liquid
Colour: Transparent (0,5 - 10 µl), yellow (2 - 200 µl and 5 - 200 µl), blue (200 - 1000 µl)
: Supplied loose or in racks holding 96 tips.
Product Family Options
- VolumeSterileTip colourPacked