VWR®, Pipette Tips, Universal
Supplier: VWR Collection
Jump to:
About this item
PP, autoclavable.
- Manufactured without lubricants
- Cadmium-free pigments
- Environmentally friendly packaging system
Colour: Yellow (2 - 200 µl), blue (50 - 1000 µl)
Length: 50 mm (2 - 200 µl), 70 mm (50 - 1000 µl)
2 Options Available Below
Product Details & Documents
PP, autoclavable.
- Manufactured without lubricants
- Cadmium-free pigments
- Environmentally friendly packaging system
Colour: Yellow (2 - 200 µl), blue (50 - 1000 µl)
Length: 50 mm (2 - 200 µl), 70 mm (50 - 1000 µl)
Documents
Product Family Options
Product Information
Accessories
Product Information
- Catalog #For use withAvailabilityPrice
- Tip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tipsTip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tips613-228850 - 1000 µl tipsLog in for availability1 * 1 items
Product Information
- Catalog #For use withAvailabilityPrice
- Tip-Box N, PP, with push on lid, stackable and autoclavable, for 200 µl tipsTip-Box N, PP, with push on lid, stackable and autoclavable, for 200 µl tips613-22872 - 200 µl tipsLog in for availability1 * 1 items