VWR®, Colour-Frosted Microscope Slides
Frosted ends in six different colours.
- Available with either cut or bevelled edges
- Writing surface resistant to all common dyes and solvents
W×D×H: 76×26×1 mm
Frosted ends in six different colours.
- Available with either cut or bevelled edges
- Writing surface resistant to all common dyes and solvents
W×D×H: 76×26×1 mm
Product Family Options
Cut edges
- DescriptionColour
Bevelled edges
- DescriptionColour