Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Disposable Micro Weighing Dishes
Supplier:   VWR Collection
VWR®, Disposable Micro Weighing Dishes
Supplier:  VWR Collection
Certificates
VWR®, Disposable Micro Weighing Dishes
VWR®, Disposable Micro Weighing Dishes
7 Options Available Below
Product Details & Documents
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
Product Family Options
Round form
Product Information
  • Capacity
    W×D×H
    Int.Ø×H
    Availability
    Price
  • 0,035 ml
    6×2,5 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • 0,35 ml
    13×3,5 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
Oval form
Product Information
  • Capacity
    W×D×H
    Int.Ø×H
    Availability
    Price
  • 0,04 ml
    6,5×3,5×2,5 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • 0,11 ml
    10×4×4,5 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • 0,12 ml
    12×2×4,5 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • 0,12 ml
    12×4×2,7 mm
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items
  • 1,12 ml
    25×7×7 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items