Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR®, Disposable Weighing Dishes with Tabs
VWR®, Disposable Weighing Dishes with Tabs
Supplier:   VWR Collection
VWR®, Disposable Weighing Dishes with Tabs
Supplier:  VWR Collection
Certificates
VWR®, Disposable Weighing Dishes with Tabs
VWR®, Disposable Weighing Dishes with Tabs
5 Options Available Below
Product Details & Documents
Aluminium.
  • Flat bottom and crimped sides for rigidity
  • Dishes feature tabs for easy handling
  • Oil-free
  • Slightly tapered for easy stacking
Sturdy, disposable dishes feature crimped sides and tabs. Ideal for general purpose weighing, dispensing and storage applications. Can also serve as evaporating dishes, parts containers or dust covers.
Product Family Options
Product Information
  • Capacity
    Int.Ø×H
    Availability
    Price
  • 6,2 ml
    28×10 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • 18,9 ml
    43×13 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 40,8 ml
    57×16 mm
    Log in for availability
    Log in for availability
    1 * 144 items
    1 * 144 items
  • 40,8 ml
    57×16 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 61,6 ml
    70×16 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items