VWR®, Disposable Weighing Dishes
Supplier: VWR Collection
Jump to:
About this item
Aluminium, light gauge.
- Flanged edge with curled lip for increased stability
- Contaminant- and oil-free
- Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
5 Options Available Below
Product Details & Documents
Aluminium, light gauge.
- Flanged edge with curled lip for increased stability
- Contaminant- and oil-free
- Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.
Documents
VWR® for Weighing
Product Literature
Product Family Options
Product Information
- Int.Ø×HAvailabilityPrice