Order Entry
Korea, Republic of
Orders LinkContactUsLinkComponent
VWR® Cooling Bricks
VWR® Cooling Bricks
Supplier:   VWR Collection
VWR® Cooling Bricks
Supplier:  VWR Collection
Certificates
VWR® Cooling Bricks
VWR® Cooling Bricks
9 Options Available Below
Product Details & Documents
VWR® cooling bricks are available in foam brick or gel filled pouch formats. The semi-rigid foam brick will holds its shape when refrigerated or frozen. The gel filled pouch can be refrigerated or frozen to keep products cold or frozen.
  • Long lasting, high-performance temperature protection
  • Suitable for pharmaceuticals, clinical trials, diagnostic kits, chemicals and biologicals
  • Available in a range of sizes
These packs are designed to safeguard temperature-sensitive products during storage and transport. They can be chilled in a freezer, refrigerator, or low temperature incubator and are effective when used with insulated shipping containers. Each cooling pack is vacuum sealed for consistent weight and shape. Phase changes at 0 °C and returns to foam state after thawing.
Product Family Options
Product Information
  • W×D×H
    Weight
    Availability
    Price
  • 216-0191
    Foam brick
    Foam brick
    114×102×19 mm
    199 g
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items
  • 216-0188
    Foam brick
    Foam brick
    114×102×38 mm
    426 g
    Log in for availability
    Log in for availability
    1 * 36 items
    1 * 36 items
  • 216-0189
    Foam brick
    Foam brick
    229×102×38 mm
    880 g
    Log in for availability
    Log in for availability
    1 * 18 items
    1 * 18 items
  • 216-0190
    Foam brick
    Foam brick
    273×236×25 mm
    1590 g
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
  • Gel pack
    165×89×19 mm
    227 g
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items
  • Gel pack
    152×140×25 mm
    341 g
    Log in for availability
    Log in for availability
    1 * 48 items
    1 * 48 items
  • Gel pack
    165×140×25 mm
    454 g
    Log in for availability
    Log in for availability
    1 * 36 items
    1 * 36 items
  • Gel pack
    203×140×32 mm
    682 g
    Log in for availability
    Log in for availability
    1 * 24 items
    1 * 24 items
  • Gel pack
    267×140×32 mm
    909 g
    Log in for availability
    Log in for availability
    1 * 18 items
    1 * 18 items