Potassium heptaiodobismuthate 0,5% in dil. acetic acid with barium chloride Reag. Ph. Eur. 1070605
Catalog # 87916.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:1253.71 g/mol
EINECS:255-595-1
CAS Number:41944-01-8
Formula:BiI₇K₄