Order Entry
Korea, Republic of
ContactUsLinkComponent
VWR®, Conductivity standard 1000 µS/cm (25 °C; 0,007 mol/L; KCl) for conductivity measurement
VWR®, Conductivity standard 1000 µS/cm (25 °C; 0,007 mol/L; KCl) for conductivity measurement
Catalog # 85642.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Conductivity standard 1000 µS/cm (25 °C; 0,007 mol/L; KCl) for conductivity measurement
Catalog # 85642.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Reference temperature:
    25 °C
  • Description:
    Conductivity standard 1000 µS/cm (0,007 mol/l; KCl)
  • Seq:
    20

Specifications

About this item

Directly traceable to NIST, tested and certified to ISO17025.