Order Entry
Korea, Republic of
ContactUsLinkComponent
Aluminium chloride Reag. Ph. Eur. 1002701
Irritant
Aluminium chloride Reag. Ph. Eur. 1002701
Catalog # 87764.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Aluminium chloride Reag. Ph. Eur. 1002701
Catalog # 87764.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :133.34 g/mol
Storage Temperature:Ambient
EINECS:231-208-1
UN:1726
ADR:8,II
 :GHS07
CAS Number:7446-70-0
Formula:AlCl₃
MDL Number:MFCD00003422