₩0
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Korea, Republic of
ContactUsLinkComponent
Oil of cedar wood
Oil of cedar wood (natural), white
Oil of cedar wood (natural), white
23686.232
:
VWR Chemicals
:
Oil of cedar wood (natural), white
23686.232
:
VWR Chemicals
:
Pack type:
Glass bottle
Pk:
250 mL
Seq:
2590
Identification:
Passes test
:
:
0.98 g/cm³ (20 °C)
:
GHS07
:
Ambient
:
68990-83-0
:
MFCD00146327