Order Entry
Korea, Republic of
ContactUsLinkComponent
 
VWR®, Propyzamide neat Certified Reference Material
  124482Q
 :  
undefined
VWR®, Propyzamide neat Certified Reference Material
  124482Q
 :  

 

  • Pack type:
    Glass ampoule
  • Pk:
    10 mg
  • CAS No.:
    23950-58-5
  • Description:
    Propyzamide neat Certified Reference Material
  • MDL No.:
    MFCD00055346
  • Formula:
    C₁₂H₁₁Cl₂NO
  • Molecular weight:
    256.13 g/mol
  • Description:
    Propyzamide