VWR® UHP Single Channel Pipette and Pipette Set
VWRU76627-792
- Colour code:Yellow
- Volume:10 - 100 µl
- Description:Single channel pipette for universal tips
- Pk:1
The VWR® UHP single channel pipette is available various sizes with a lightweight and durable design. These pipettes will provide you with low pipetting forces and the highest levels of accuracy and precision.
- Autolocking volume mechanism to avoid volume drift while pipetting
- Fully autoclaveable and UV resistant
- Color coded ejector button and calibration switch make for easy identification
- Universal fit with most common brands of pipette tips
- Lightweight design, quality materials, low pipetting forces, and a 5-year warranty
These single channel pipettes are calibrated to ISO 8655 standards using VWR® Universal pipette tips. Pipettes were tested for both autoclave and UV resistance.
: In compliance with ISO 8655.
: Includes a QC certificate, pipette, quick reference guide, and calibration tool.
Pipette sets include the pipettes, a rack of each compatible pipette tip, QC certificate and calibration tools.
Pipette sets include the pipettes, a rack of each compatible pipette tip, QC certificate and calibration tools.