VWR® Short Thread Vials and Septa, Wide Opening, ND9
Short screw thread ND9, wide opening, thread HPLC vial with matched PP screw caps ND9, PTFE/silicone septa and micro-insert.
- Screw vials can be used on all common autosampler
- Short screw, long neck to make the autosampler easier to capture
- Wider opening for easy puncture
- PTFE/silicone septa, has excellent chemical inertness, acid and alkali resistance
- Screw blue open top pp caps
Short screw thread ND9 vial is designed to ensure proper centering of limited volume inserts, eliminating missed injections. Available with black or blue screw caps with 9 mm thread. Available as closed top screw seals or with centre hole. Vials with integrated micro-insert are also available.
Short screw thread ND9 vial is designed to ensure proper centering of limited volume inserts, eliminating missed injections. Vials have a 40% larger opening for easier sample preparation.
: The combopacks of vials and caps contain vials and septa caps - 100 pieces each.