


Calcium sulphate, Drierite™ (4 Mesh), with indicator (cobalt (II) chloride), J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7778-18-9
:
Formula:CaO₄S
MDL Number:MFCD00010912
:136.14 g/mol
EINECS:231-900-3
:GHS08
:GHS06
:GHS09
Storage Temperature:Ambient
Frequently Bought Together