Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
₩0
주문 입력
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
크로마토그래피 레진
HPLC Sorbents
Octadecyl (C18), PolarPlus®
Octadecyl (C18), PolarPlus®
Octadecyl (C18), PolarPlus®
Catalog # 7204-R
Supplier:
Avantor
CAS Number:
Certificates
이미지 확대
Octadecyl (C18), PolarPlus®
Catalog # 7204-R
Supplier:
Avantor
CAS Number:
Certificates
Specifications
입자 크기:
10 µm
Pk:
1 g
설명:
PolarPlus® octadecyl (C18)
Specifications
About this item
Product Details
Documentation
Product Details
Documentation