



Hexane (mixture of isomers), low water content ≥98.5% (by GC), BakerDRY™
Supplier: Avantor
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Density:0.66 g/cm³
Flash Pt:–23.00 °C
UN:1208
ADR:3,II
:Management of Change (MOC) category = NI
:GHS02
:GHS07
:GHS08
:GHS09
Specification Test Results
- ACS - Assay (C₆H₁₄) (by GC)(total isomers):≥98.5 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤2 ppm
- ACS - Water-Soluble Titrable Acid, meq/g:≤0.0003
- ACS - Sulfur Compounds (as S):≤0.005 %
- ACS - Thiophene:Passes Test
- ACS - Water (by KF, coulometric):≤20 ppm
Specification Test Results
Frequently Bought Together