Molecular sieve 3A (0.3 nm, 3 Å), rods, GPR RECTAPUR®, Ø ~1.6 mm
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:308080-99-1
:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613
Storage Temperature:Ambient
EINECS:215-283-8
Specification Test Results
- Identification:Passes test
- Loss on ignition (575 °C):max. 2.0%
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC28461.230 spec_VWRC28461.292
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together