0
₩0
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Korea, Republic of
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Centrifuges
Rotors
VWR® Mini 12, PCR Strip Rotor for Mini Centrifuge
VWR® Mini 12, PCR Strip Rotor for Mini Centrifuge
VWR® Mini 12, PCR Strip Rotor for Mini Centrifuge
Catalog # VWRA521-2796
Supplier:
VWR Collection
Expand image
VWR® Mini 12, PCR Strip Rotor for Mini Centrifuge
Catalog # VWRA521-2796
Supplier:
VWR Collection
Specifications
Description:
PCR strip rotor with 2×8×0,2 ml capacity
Pk:
1
Specifications
About this item
Product Details
Documentation
Product Details
Documentation