


Alcohol, anhydrous, SpectrAR, Macron Fine Chemicals™
Catalog # MACR9919-88
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v):94.0 - 96.0 %
- Isopropyl Alcohol (2-propanol)(v/v):4.0 - 6.0 %
- Ethanol (v/v):88.5 - 92.5 %
- Methanol (v/v):3.5 - 5.5 %
- Appearance:Passes Test
- Color (APHA):≤ 5
- Residue after Evaporation:≤ 5 ppm
- Specific Gravity at 25°C:0.785 - 0.795
- Water (H₂O):≤ 0.1 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 210 nm:≤ 1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 220 nm:≤ 0.50
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 230 nm:≤ 0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 250 nm:≤ 0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 270 nm:≤ 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400 nm:≤ 0.01
- Substances Reducing Permanganate:Passes Test
Specification Test Results
About this item
:
Storage Temperature:Ambient
EINECS:200-578-6
UN:1987
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:64-17-5
MDL Number:MFCD00003568