

Dichloromethane ≥99.8% (by GC, corrected for water content) stabilised for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
9315-88
: Avantor
:
- Pk:4 L
- Seq:5162
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:≤ 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤ 233
- Assay (by GC, corrected for water, exclusive of preservative):≥ 99.8 %
- Chloride (Cl):≤ 10 ppm
- Residue after Evaporation:≤ 2.0 ppm
- Titrable Acid (µeq/g):≤ 0.3
- Water (by KF, coulometric):≤ 0.02 %
- Preservative (Amylene):≤ 200 ppm
:
:84.93 g/mol
:12,06140
:39.8 °C (1013 hPa)
:–95 °C
:1.322 g/cm³ (20 °C)
:Ambient
:6.1,III
:Packaged under nitrogen.
:Stabilised with amylene <= 200 ppm
:200-838-9
:1593
:GHS08
:GHS07
:75-09-2
:CH₂Cl₂
:MFCD00000881