4-Aminobutyric acid ≥99%, Biotech Reagent, J.T.Baker®
Catalog # V265-10
Supplier: Avantor
CAS Number:
Specifications
- Pk:10,2 kg
- Seq:4169
- Pk Info:1×10 kg
Specifications
Specification Test Results
- Appearance (White to off-white solid):Passes Test
- Identification (FTIR):Passes Test
- Clarity of solution (5% in water):Passes Test
- Assay:≥99 %
- Loss on drying (105°C):≤0.5 %
- Residual solvent - Methanol:≤3000 ppm
- Residual solvent - Ethanol:≤5000 ppm
- Residual solvent - Isopropanol:≤5000 ppm
- Aluminum (Al):≤60 ppm
- Cadmium (Cd):≤2 ppm
- Chromium (Cr):≤7 ppm
- Cobalt (Co):≤100 ppm
- Copper (Cu):≤20 ppm
- Manganese (Mn):≤10 ppm
- Zinc (Zn):≤900 ppm
- Nickel (Ni):≤20 ppm
- Selenium (Se):≤25 ppm
Specification Test Results
About this item
:
:103.12 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:195…204 °C
:Management of Change (MOC) category = R
EINECS:200-258-6
CAS Number:56-12-2
Formula:C₄H₉NO₂
MDL Number:MFCD00008226