Order Entry
Korea, Republic of
ContactUsLinkComponent
Nitric acid 7.7 N, ULTREX® II, Ultrapure, J.T.Baker®
Oxidizing
Corrosive
Irritant
Nitric acid 7.7 N, ULTREX® II, Ultrapure, J.T.Baker®
Catalog # 6985-05
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Corrosive
Irritant
Nitric acid 7.7 N, ULTREX® II, Ultrapure, J.T.Baker®
Catalog # 6985-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725
  • Pk Info:
    1×500 ml

Specifications

Specification Test Results

  • Normality:
    7.5 - 7.9
  • Trace Impurities - Aluminum (Al):
    ≤100 ppt
  • Trace Impurities - Antimony (Sb):
    ≤100 ppt
  • Trace Impurities - Arsenic (As):
    ≤100 ppt
  • Trace Impurities - Barium (Ba):
    ≤100 ppt
  • Trace Impurities - Beryllium (Be):
    ≤100 ppt
  • Trace Impurities - Bismuth (Bi):
    ≤100 ppt
  • Trace Impurities - Boron (B):
    ≤300 ppt
  • Trace Impurities - Cadmium (Cd):
    ≤100 ppt
  • Trace Impurities - Calcium (Ca):
    ≤100 ppt
  • Trace Impurities - Cerium (Ce):
    ≤20 ppt
  • Trace Impurities - Cesium (Cs):
    ≤20 ppt
  • Trace Impurities - Chromium (Cr):
    ≤100 ppt
  • Trace Impurities - Cobalt (Co):
    ≤100 ppt
  • Trace Impurities - Copper (Cu):
    ≤100 ppt
  • Trace Impurities - Dysprosium (Dy):
    ≤20 ppt
  • Trace Impurities - Erbium (Er):
    ≤20 ppt
  • Trace Impurities - Europium (Eu):
    ≤20 ppt
  • Trace Impurities - Gadolinium (Gd):
    ≤20 ppt
  • Trace Impurities - Gallium (Ga):
    ≤20.0 ppt
  • Trace Impurities - Germanium (Ge):
    ≤100 ppt
  • Trace Impurities - Gold (Au):
    ≤100 ppt
  • Trace Impurities - Holmium (Ho):
    ≤20 ppt
  • Trace Impurities - Indium (In):
    ≤20 ppt
  • Trace Impurities - Iron (Fe):
    ≤100 ppt
  • Trace Impurities - Lanthanum (La):
    ≤20 ppt
  • Trace Impurities - Lead (Pb):
    ≤10 ppt
  • Trace Impurities - Lithium (Li):
    ≤20 ppt
  • Trace Impurities - Lutetium (Lu):
    ≤20 ppt
  • Trace Impurities - Magnesium (Mg):
    ≤100 ppt
  • Trace Impurities - Manganese (Mn):
    ≤100 ppt
  • Trace Impurities - Mercury (Hg):
    ≤100 ppt
  • Trace Impurities - Molybdenum (Mo):
    ≤100 ppt
  • Trace Impurities - Neodymium (Nd):
    ≤20 ppt
  • Trace Impurities - Nickel (Ni):
    ≤100 ppt
  • Trace Impurities - Niobium (Nb):
    ≤20 ppt
  • Trace Impurities - Palladium (Pd):
    ≤20 ppt
  • Trace Impurities - Platinum (Pt):
    ≤100 ppt
  • Trace Impurities - Potassium (K):
    ≤100 ppt
  • Trace Impurities - Praseodymium (Pr):
    ≤20 ppt
  • Trace Impurities - Rhodium (Rh):
    ≤20 ppt
  • Trace Impurities - Rubidium (Rb):
    ≤20 ppt
  • Trace Impurities - Samarium (Sm):
    ≤20 ppt
  • Trace Impurities - Scandium (Sc):
    ≤20 ppt
  • Trace Impurities - Silver (Ag):
    ≤10 ppt
  • Trace Impurities - Sodium (Na):
    ≤100 ppt
  • Trace Impurities - Strontium (Sr):
    ≤10 ppt
  • Trace Impurities - Terbium (Tb):
    ≤20 ppt
  • Trace Impurities - Thallium (Tl):
    ≤10 ppt
  • Trace Impurities - Thorium (Th):
    ≤1 ppt
  • Trace Impurities - Thulium (Tm):
    ≤20 ppt
  • Trace Impurities - Tin (Sn):
    ≤100 ppt
  • Trace Impurities - Titanium (Ti):
    ≤100 ppt
  • Trace Impurities - Tungsten (W):
    ≤100 ppt
  • Trace Impurities - Uranium (U):
    ≤1 ppt
  • Trace Impurities - Vanadium (V):
    ≤100 ppt
  • Trace Impurities - Ytterbium (Yb):
    ≤20 ppt
  • Trace Impurities - Zinc (Zn):
    ≤100 ppt
  • Trace Impurities - Zirconium (Zr):
    ≤10 ppt

Specification Test Results

About this item

 :
 :63.01 g/mol
ADR:8,II
 :Management of Change (MOC) category = N
UN:2031
 :GHS03
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349