


Toluene ≥99.7% (by GC, corrected for water content), ChromAR®, Macron Fine Chemicals™
Catalog # MACR9918-88
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Assay (by GC, corrected for water):≥ 99.7 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤ 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:≤ 0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm:≤ 0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤ 285
- Residue after Evaporation:≤ 3.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤ 0.02 %
Specification Test Results
About this item
:
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.8636 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1294
ADR:3,II
:Management of Change (MOC) category = N
REACH:05-2114615130-69
:GHS02
:GHS08
:GHS07
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512