

Dichloromethane ≥99.8% (without stabilizer, GC), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9916-88
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤ 5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤ 10
- Assay (CH₂Cl₂) (by GC, exclusive of preservative, corrected for water):≥ 99.8 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 1.0 ppm
- Titrable Acid (µeq/g):≤ 0.3
- Chloride (Cl):≤ 10 ppm
- Water (by KF, coulometric):≤ 0.02 %
Specification Test Results
About this item
:
:84.93 g/mol
Boiling Pt:39…40 °C
Melting Pt:–95 °C
Storage Temperature:Ambient
ADR:6.1,III
:Management of Change (MOC) category = NI
UN:1593
:GHS07
:GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881