Order Entry
Korea, Republic of
ContactUsLinkComponent
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (95% n-hexane), UltimAR®, Macron Fine Chemicals™
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (95% n-hexane), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9915-88
CAS Number:  
Flammable
Health
Irritant
Environment
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (95% n-hexane), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9915-88
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤ 5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤ 10
  • Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):
    ≥ 99.5 %
  • Assay (as n-Hexane) (by GC, corrected for water):
    ≥ 95 %
  • Color (APHA):
    ≤ 10
  • Residue after Evaporation:
    ≤ 1.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤ 0.05 %

Specification Test Results

About this item

 :
 :86.18 g/mol
Melting Pt:–95 °C
UN:1208
ADR:3,II
 :Management of Change (MOC) category = NI
Flash Pt:–23 °C
 :GHS02
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311