


Ethyl acetate ≥99.6% (by GC, corrected for water content), UltimAR®, Macron Fine Chemicals™
Catalog # MACR9914-88
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤ 5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤ 10
- Assay (CH₃COOC₂H₅) (by GC, corrected for water):≥ 99.6 %
- Titrable Acid (µeq/g):≤ 0.8
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 1.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, volumetric):≤ 0.05 %
Specification Test Results
About this item
:
:88.11 g/mol
Merck Index:13,03792
Melting Pt:–83 °C
UN:1173
ADR:3,II
:Management of Change (MOC) category = N
Flash Pt:–4 °C
EINECS:205-500-4
REACH:01-2119475103-46
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:141-78-6
Formula:CH₃COOC₂H₅
MDL Number:MFCD00009171