Order Entry
Korea, Republic of
ContactUsLinkComponent
 
VWR®, Disposal Carton for Plastics
  VWRU58949-700
undefined
VWR®, Disposal Carton for Plastics
  VWRU58949-700

 

  • Capacity:
    38 L
  • Description:
    Disposal box
  • W×D×H:
    305×305×686 mm
  • Pk:
    6

 

 

Disposal boxes are shipped flat and fold into shape easily.

  • Clearly labelled, rigid cardboard carton; includes polypropylene liner to contain spills
  • Dedicated to separating plastics from the waste stream
  • Sized to fit under many benchtops
  • Approximate volume: 38 L

Entire box can be discarded when filled, ensuring safe handling and ready identification of contents